Skip to Content
Merck
All Photos(1)

Documents

739340

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate)

0.54% in H2O, high-conductivity grade

Synonym(s):

Orgacon S305, PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

grade

high-conductivity grade

form

liquid

contains

1-5% Propan-2-ol
5-10% Diethylene glycol

concentration

0.54% in H2O

resistance

<200 Ω/sq, >90% visible light transmission (40 μm wet)

refractive index

n20/D 1.344

pH

<2.7

viscosity

<18 cP(22 °C)

density

0.983 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

A conducting polymer such as poly(3,4-ethylenedioxythiophene) doped with poly(styrene sulfonate) anions (PEDOT/PSS) is widely used in various organic optoelectronic devices. PEDOT: PSS is a blend of cationic polythiopene derivative, doped with a polyanion. High electrical conductivityand good oxidation resistance of such polymers make it suitable for electromagnetic shielding and noise suppression. Thus, the polymer film was found to possess high transparency throughout the visible light spectrum and even into near IR and near UV regions, virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Impact of small electric and magnetic fields on the polymer was studied.

Application

Aqueous formulation of high conductivity grade PEDOT:PSS polymer dispersion suitable for transparent conductive coatings. Includes PEDOT:PSS and surfactants.
Used in ITO-free OPV applications as a highly conductive hole transport layer. Also can be used for optical films, touch pads, touch screen displays, flexible LCDs, and e-paper applications.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Legal Information

Product of Agfa-Gevaert N.V.
Orgacon is a trademark of Agfa-Gevaert N.V.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metal salt modified PEDOT:PSS as anode buffer layer and its effect on power conversion efficiency of organic solar cells
Kadem B, et al.
Organic Electronics, 24, 73-79 (2015)
Simple casting based fabrication of PEDOT:PSS-PVDF-ionic liquid soft actuators
Simaite A, et al.
SPIE Proc. null
Solar cells made from polymers containing Dithieno [3, 2-b: 2', 3'-d] pyrrole with different side chain lengths
Gong C, et al.
Solar Energy Mat. and Solar Cells, 95(3), 969-973 (2011)
Mou Pal et al.
Nanoscale research letters, 7(1), 1-1 (2012-01-05)
Uniform, spherical-shaped TiO2:Eu nanoparticles with different doping concentrations have been synthesized through controlled hydrolysis of titanium tetrabutoxide under appropriate pH and temperature in the presence of EuCl3·6H2O. Through air annealing at 500°C for 2 h, the amorphous, as-grown nanoparticles could
SURPRISING IMPACT OF SMALL ELECTRIC AND MAGNETIC FIELDS ON CONDUCTING POLYMERS
Epstein AJ, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 48(1), 119-119 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service