Skip to Content
Merck
All Photos(1)

Key Documents

738158

Sigma-Aldrich

N-(Allyloxycarbonyloxy)succinimide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO5
CAS Number:
Molecular Weight:
199.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.482

density

1.287 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=CCOC(=O)ON1C(=O)CCC1=O

InChI

1S/C8H9NO5/c1-2-5-13-8(12)14-9-6(10)3-4-7(9)11/h2H,1,3-5H2

InChI key

OIXALTPBNZNFLJ-UHFFFAOYSA-N

Application

N-(Allyloxycarbonyloxy)succinimide (alloc-Su) can be used as:
  • A building block for the preparation of glycopeptide scaffolds.[1]
  • A reagent in the synthesis of various functional cyclic carbonate monomers from 2-amino-1,3-propane diols.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

242.6 °F

Flash Point(C)

117 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and in vitro evaluation of bisphosphonated glycopeptide prodrugs for the treatment of osteomyelitis
Tanaka KSE, et al.
Bioorganic & medicinal chemistry letters, 20(4), 1355-1359 (2010)
2-Amino-1, 3-propane diols: a versatile platform for the synthesis of aliphatic cyclic carbonate monomers
Venkataraman S, et al.
Polym. Chem., 4(10), 2945-2948 (2013)
Bo Wu et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-07)
Four bis-lactam [i, i+4]-stapled peptides with d- or l-α-methyl-thialysines were constructed on a model peptide sequence derived from p110α[E545K] and subjected to circular dichroism (CD) and proteolytic stability assessment, alongside the corresponding bis-lactam [i, i+4]-stapled peptide with l-thialysine. The %

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service