Skip to Content
Merck
All Photos(1)

Key Documents

737593

Sigma-Aldrich

2-(4-Aminophenyl)benzothiazole

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€70.90

€70.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€70.90

About This Item

Empirical Formula (Hill Notation):
C13H10N2S
CAS Number:
Molecular Weight:
226.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€70.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

151-156 °C

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)-c2nc3ccccc3s2

InChI

1S/C13H10N2S/c14-10-7-5-9(6-8-10)13-15-11-3-1-2-4-12(11)16-13/h1-8H,14H2

InChI key

WKRCOZSCENDENK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stephan Mokesch et al.
Frontiers in chemistry, 8, 209-209 (2020-04-23)
A series of 2-phenylbenzothiazole derivatives and their corresponding organometallic ruthenium(II) and osmium(II) complexes were synthesized, designed to exploit both, the attributes of the half-sandwich transition metal scaffold and the bioactivity spectrum of the applied 2-phenylbenzothiazoles. All synthesized compounds were characterized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service