Skip to Content
Merck
All Photos(1)

Documents

737550

Sigma-Aldrich

Ethyl imidazo[1,2-a]pyridine-2-carboxylate

97%

Synonym(s):

Imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O2
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

83-87 °C

SMILES string

CCOC(=O)c1cn2ccccc2n1

InChI

1S/C10H10N2O2/c1-2-14-10(13)8-7-12-6-4-3-5-9(12)11-8/h3-7H,2H2,1H3

InChI key

GNFACXDTRBVZJE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service