Skip to Content
Merck
All Photos(1)

Key Documents

736635

Sigma-Aldrich

Bis(tricyclohexylphosphine)palladium(0)

Synonym(s):

Pd(PCy3)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H66P2Pd
CAS Number:
Molecular Weight:
667.28
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

87-92 °C

storage temp.

2-8°C

SMILES string

[Pd].C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C4CCC(CC4)P(C5CCCCC5)C6CCCCC6

InChI

1S/2C18H33P.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h2*16-18H,1-15H2;

InChI key

JGBZTJWQMWZVNX-UHFFFAOYSA-N

Application

Bis(tricyclohexylphosphine)palladium(0) [Pd(PCy3)2] is a general palladium precatalyst that can be used to catalyze various reactions such as thioesterification of alkynes, alumination of unactivated arenes, direct arylation polycondensation and intramolecular alkane arylation adjacent to amides and sulfonamides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct arylation polycondensation for synthesis of optoelectronic materials.
Kuwabara J.
Polymer Journal (2018)
Palladium-catalyzed thioesterification of alkynes with O-methyl S-phenyl thiocarbonate.
Hua R, et al.
Journal of the American Chemical Society, 123(12), 2899-2900 (2001)
Investigation of the mechanism of C (sp3)? H bond cleavage in Pd (0)-catalyzed intramolecular alkane arylation adjacent to amides and sulfonamides.
Rousseaux S, et al.
Journal of the American Chemical Society, 132(31), 10692-10705 (2010)
Room temperature catalytic carbon?hydrogen bond alumination of unactivated arenes: mechanism and selectivity.
Hooper T, et al.
Chemical Science, 9, 5435-5440 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service