Skip to Content
Merck
All Photos(2)

Key Documents

736589

Sigma-Aldrich

2,6-Bis(di-tert-butylphosphinomethyl)pyridine

Synonym(s):

2,6-Bis(di-t-butylphosphinomethyl)pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€144.00
1 G
€294.00

€144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€144.00
1 G
€294.00

About This Item

Empirical Formula (Hill Notation):
C23H43NP2
CAS Number:
Molecular Weight:
395.54
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€144.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

116-119 °C

functional group

phosphine

SMILES string

CC(C)(C)P(Cc1cccc(CP(C(C)(C)C)C(C)(C)C)n1)C(C)(C)C

InChI

1S/C23H43NP2/c1-20(2,3)25(21(4,5)6)16-18-14-13-15-19(24-18)17-26(22(7,8)9)23(10,11)12/h13-15H,16-17H2,1-12H3

InChI key

NJQAVBPPWNSBBC-UHFFFAOYSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

154.0 °F

Flash Point(C)

67.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service