Skip to Content
Merck
All Photos(1)

Key Documents

736406

Sigma-Aldrich

4-(4-Iodophenyl)morpholine

97%

Synonym(s):

4-Iodomorpholinobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€129.00

About This Item

Empirical Formula (Hill Notation):
C10H12INO
CAS Number:
Molecular Weight:
289.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

158-163 °C

functional group

ether
iodo

SMILES string

Ic1ccc(cc1)N2CCOCC2

InChI

1S/C10H12INO/c11-9-1-3-10(4-2-9)12-5-7-13-8-6-12/h1-4H,5-8H2

InChI key

YUEREUIIUISPFV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service