Skip to Content
Merck
All Photos(1)

Key Documents

735329

Sigma-Aldrich

(Iodomethyl)cyclopentane

contains copper as stabilizer, 95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€161.00

About This Item

Empirical Formula (Hill Notation):
C6H11I
CAS Number:
Molecular Weight:
210.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€161.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.537

density

1.604 g/mL at 25 °C

functional group

iodo

SMILES string

ICC1CCCC1

InChI

1S/C6H11I/c7-5-6-3-1-2-4-6/h6H,1-5H2

InChI key

DUMSKQUKLVSSII-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Asp. Tox. 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service