Skip to Content
Merck
All Photos(1)

Key Documents

733776

Sigma-Aldrich

Trimethyl phosphite solution

NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€423.00

€423.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 PKG
€423.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.12

€423.00


Check Cart for Availability

Request more information

grade

NMR reference standard

Quality Level

form

solution

concentration

1% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

10 mm × 8 in.

suitability

suitable for NMR (reference standard)

SMILES string

COP(OC)OC

InChI

1S/C3H9O3P/c1-4-7(5-2)6-3/h1-3H3

InChI key

CYTQBVOFDCPGCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

31P sensitivity

Quantity

10 mm O.D. tube contains 3.175 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service