Skip to Content
Merck
All Photos(2)

Key Documents

733261

Sigma-Aldrich

4-(Difluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€76.10

€76.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€76.10

About This Item

Empirical Formula (Hill Notation):
C6H5F2N
CAS Number:
Molecular Weight:
129.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€76.10


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.463

density

1.207 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)c1ccncc1

InChI

1S/C6H5F2N/c7-6(8)5-1-3-9-4-2-5/h1-4,6H

InChI key

XPNKTOPRNGWNCL-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

124.0 °F

Flash Point(C)

51.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service