Skip to Content
Merck
All Photos(1)

Key Documents

732656

Sigma-Aldrich

Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate

Synonym(s):

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O2
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

42-46 °C

functional group

ester

SMILES string

CCOC(=O)c1cc(C)n(C)n1

InChI

1S/C8H12N2O2/c1-4-12-8(11)7-5-6(2)10(3)9-7/h5H,4H2,1-3H3

InChI key

OJPXVXXMBWKEAT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service