Skip to Content
Merck
All Photos(1)

Key Documents

73200

Sigma-Aldrich

O-(4-Nitrobenzyl)hydroxylamine hydrochloride

≥98.5% (AT)

Synonym(s):

4-Nitrobenzyloxyamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2ONH2 · HCl
CAS Number:
Molecular Weight:
204.61
Beilstein:
3709565
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5% (AT)

form

crystals

mp

215 °C (dec.) (lit.)

functional group

nitro

SMILES string

Cl.NOCc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H8N2O3.ClH/c8-12-5-6-1-3-7(4-2-6)9(10)11;/h1-4H,5,8H2;1H

InChI key

LKCAFSOYOMFQSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the preparation of N-(4-nitrobenzyloxy)-amino acids as substrates for an unambiguous N-hydroxypeptide synthesis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth P Roberts et al.
Chemical research in toxicology, 19(2), 300-309 (2006-02-21)
A new method has been developed to accurately measure apurinic and apyrimidinic (AP) DNA damage sites, which are lesions in DNA formed by loss of a nucleobase from oxidative stress or carcinogen adducts. If AP sites are left unrepaired (or
M Pauly et al.
Planta, 212(5-6), 842-850 (2001-05-12)
Xyloglucans were isolated by sequential extraction of the cell walls of pea (Pisum sativum L. cv. Alaska) with a xyloglucan-specific endoglucanase and KOH. The xyloglucan content and xyloglucan-oligosaccharide composition were determined for fractions obtained from the elongating and non-elongating segments
B X Chen et al.
Mutation research, 273(3), 253-261 (1992-05-01)
The abasic site is one of the most frequent changes occurring in DNA and has been shown to be lethal and mutagenic. An abasic site in DNA can be tagged by reaction with O-4-nitrobenzylhydroxylamine (NBHA), resulting in the formation of
M Pauly et al.
Carbohydrate research, 282(1), 1-12 (1996-02-28)
An improved procedure has been developed for the rapid derivatization of oligosaccharides with UV-detectable p-nitrobenzylhydroxylamine (PNB). The improved conditions used result in quantitative derivatization of neutral oligosaccharides. Sialylated oligosaccharides can also be quantitatively PNB-derivatized without detectable desialylation. Of the oligosaccharides
Joo Hyun Lim et al.
Frontiers in microbiology, 9, 2333-2333 (2018-10-16)
2-Deoxy-scyllo-inosose (DOI) has been a valuable starting natural product for the manufacture of pharmaceuticals or chemical engineering resources such as pyranose catechol. DOI synthase, which uses glucose-6-phosphate (Glc6P) as a substrate for DOI biosynthesis, is indispensably involved in the initial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service