Skip to Content
Merck
All Photos(1)

Key Documents

731544

Sigma-Aldrich

2-Oxa-7-azaspiro[3.5]nonane oxalate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€199.00

€199.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€199.00

About This Item

Empirical Formula (Hill Notation):
C8H14NO3 · xC2H2O4
CAS Number:
Molecular Weight:
172.20 (free base basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€199.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

functional group

carboxylic acid
ether

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.C1CC2(CCN1)COC2

InChI

1S/C7H13NO.C2H2O4/c1-3-8-4-2-7(1)5-9-6-7;3-1(4)2(5)6/h8H,1-6H2;(H,3,4)(H,5,6)

InChI key

KFMUWBFXUTUOEX-UHFFFAOYSA-N

Other Notes

PRODUCT IS NOT AVAILABLE IN EUROPE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayinuer Maitituoheti et al.
Cell reports, 33(3), 108293-108293 (2020-10-22)
Histone methyltransferase KMT2D harbors frequent loss-of-function somatic point mutations in several tumor types, including melanoma. Here, we identify KMT2D as a potent tumor suppressor in melanoma through an in vivo epigenome-focused pooled RNAi screen and confirm the finding by using a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service