Skip to Content
Merck
All Photos(1)

Documents

731250

Sigma-Aldrich

Tris[2-(3-mercaptopropionyloxy)ethyl] isocyanurate

Synonym(s):

3TI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27N3O9S3
CAS Number:
Molecular Weight:
525.62
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.538

density

1.333 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

SCCC(=O)OCCN1C(=O)N(CCOC(=O)CCS)C(=O)N(CCOC(=O)CCS)C1=O

InChI

1S/C18H27N3O9S3/c22-13(1-10-31)28-7-4-19-16(25)20(5-8-29-14(23)2-11-32)18(27)21(17(19)26)6-9-30-15(24)3-12-33/h31-33H,1-12H2

InChI key

CFKONAWMNQERAG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamil Charubin et al.
mBio, 11(5) (2020-09-03)
Microbial syntrophy is universal in nature, profoundly affecting the composition and function of microbiomes. We have recently reported data suggesting direct cell-to-cell interactions leading to electron and material exchange between the two microbes in the syntrophy between Clostridium ljungdahlii and
Ang Li et al.
Bioresource technology, 305, 123066-123066 (2020-03-03)
Clostridium diolis can efficiently utilize various inexpensive, renewable resources such as crude glycerol and lignocellulosic biomass hydrolysate to produce bulk chemicals and fuels. However, its study has been impeded by the lack of efficient plasmids electro-transformation techniques. In this study
Zhiqiang Wen et al.
Applied and environmental microbiology, 85(7) (2019-01-20)
Clostridium cellulovorans DSM 743B offers potential as a chassis strain for biomass refining by consolidated bioprocessing (CBP). However, its n-butanol production from lignocellulosic biomass has yet to be demonstrated. This study demonstrates the construction of a coenzyme A (CoA)-dependent acetone-butanol-ethanol
Hoyle, C.; Lee, Tai Yeon; Roper, Todd
Journal of Polymer Science Part A: Polymer Chemistry, 42 (2004)
Lauren A Riley et al.
Biotechnology for biofuels, 14(1), 30-30 (2021-01-27)
Non-model microorganisms often possess complex phenotypes that could be important for the future of biofuel and chemical production. They have received significant interest the last several years, but advancement is still slow due to the lack of a robust genetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service