Skip to Content
Merck
All Photos(3)

Key Documents

730777

Sigma-Aldrich

Silver, dispersion

nanoparticles, 100 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer

Synonym(s):

Colloidal silver

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€152.00

€152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€152.00

About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

€152.00


Check Cart for Availability

Request a Bulk Order

form

nanoparticles

contains

sodium citrate as stabilizer

concentration

0.02 mg/mL in aqueous buffer

refractive index

n20/D 1.333

particle size

100 nm (TEM)

density

0.9978 g/mL at 25 °C

fluorescence

λem 504 nm FWHM 167 nm

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The silver nanoparticles with a particle size of 100nm and refractive index of 1.33, have unique properties such as high electronic conductivity, a large surface area, and a narrow fluorescence spectrum, making them suitable for a range of applications. They are widely used in the field of drug delivery systems, imaging probes, diagnostic platforms, and energy storage devices.

Application

  • Silver as acrolein hydrogenation catalyst: intricate effects of catalyst nature and reactant partial pressures.: This study explores the use of silver as a catalyst for acrolein hydrogenation, highlighting the complex effects of catalyst nature and the partial pressures of reactants, which could inform future applications in industrial chemical processes (Bron et al., 2007).
  • Surface-enhanced Raman analysis of sulfa drugs on colloidal silver dispersion.: This research discusses the application of surface-enhanced Raman spectroscopy (SERS) using colloidal silver dispersions for analyzing sulfa drugs, potentially enhancing detection methods in pharmaceutical research (Sutherland et al., 1990).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ebtehal Ahmed et al.
Journal of biomedical materials research. Part A, 108(12), 2351-2367 (2020-05-18)
Acute liver failure (ALF) occurs due to severe liver damage that triggers rapid loss of normal liver function. Here, we investigate the usefulness of an injectable liver extracellular matrix (LECM)-rich hydrogel generated from an optimized decellularization protocol incorporated with silver

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service