Skip to Content
Merck
All Photos(1)

Documents

729949

Sigma-Aldrich

1-Bromo-4-(1,1-difluoroethyl)benzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrF2
CAS Number:
Molecular Weight:
221.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.503

density

1.495 g/mL at 25 °C

SMILES string

CC(F)(F)c1ccc(Br)cc1

InChI

1S/C8H7BrF2/c1-8(10,11)6-2-4-7(9)5-3-6/h2-5H,1H3

InChI key

QXIBKCFAFRHORF-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

180.0 °F

Flash Point(C)

82.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service