Skip to Content
Merck
All Photos(1)

Documents

728799

Sigma-Aldrich

Potassium 2-bromopyridine-3-trifluoroborate

97%

Synonym(s):

Potassium 2-bromopyridin-3-yl-trifluoroborate, Potassium(2-bromopyridin-3-yl)trifluoroborate, Potassium-2-bromo-3-pyridyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BBrF3KN
Molecular Weight:
263.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

250-255 °C

SMILES string

[K+].F[B-](F)(F)c1cccnc1Br

InChI

1S/C5H3BBrF3N.K/c7-5-4(6(8,9)10)2-1-3-11-5;/h1-3H;/q-1;+1

InChI key

HEDPVDPUNQXJGE-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service