Skip to Content
Merck
All Photos(1)

Key Documents

728640

Sigma-Aldrich

11-Mercaptoundecylhydroquinone

95%

Synonym(s):

2-(11-Mercaptoundecyl)-1,4-benzenediol, MUH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28O2S
CAS Number:
Molecular Weight:
296.47
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

mp

86-92 °C

storage temp.

−20°C

SMILES string

Oc1ccc(O)c(CCCCCCCCCCCS)c1

InChI

1S/C17H28O2S/c18-16-11-12-17(19)15(14-16)10-8-6-4-2-1-3-5-7-9-13-20/h11-12,14,18-20H,1-10,13H2

InChI key

DYYJRXGKNZGOIX-UHFFFAOYSA-N

Application

11-Mercaptoundecylhydroquinone forms a thiol based self-assembled monolayer (SAM) that facilitates the surface modification by immobilizing the surface atoms. It can be used in functionalization of quantum dots for potential application in sensors and other optoelectronics devices. It can also be coated on the gold electrode for electrical energy storage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy A Rigotti et al.
Trials, 21(1), 336-336 (2020-04-18)
Tobacco smoking remains the leading preventable cause of death in the US. A hospital admission provides smokers with a unique opportunity to stop smoking because it requires temporary tobacco abstinence while illness may enhance motivation to quit. Hospital interventions must
Directed Assembly of Quantum Dots Using Brush Block Copolymers for Well-Ordered Nonlinear Optical Nanocomposites
Song DP, et al.
Macromolecules, 49(14), 5068-5075 (2016)
Christophe Millien et al.
International journal for equity in health, 20(1), 1-1 (2021-01-03)
Uterine fibroids, the most common cause of gynecologic surgery, have a reported cumulative incidence of 59% among Black women in the U.S. Uterine fibroids negatively impact the quality of women's lives. No study has been found in the literature about
Travis D Satterlund et al.
Tobacco control, 23(6), 491-495 (2013-06-21)
Although it is legal for multiunit housing (MUH) property owners in all 50 states to prohibit smoking on their premises, including in individual units, MUH constitutes a relatively new setting to reduce exposure to secondhand smoke via voluntary smoke-free policy.
Sherif T S Hassan
Biomedicines, 8(5) (2020-05-28)
While few studies have revealed the biological properties of brassicasterol, a phytosterol, against some biological and molecular targets, it is believed that there are still many activities yet to be studied. In this work, brassicasterol exerts a therapeutic utility in

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service