Skip to Content
Merck
All Photos(1)

Key Documents

728462

Sigma-Aldrich

PyOxim

≥97.5% (HPLC), for peptide synthesis

Synonym(s):

[Ethyl cyano(hydroxyimino)acetato-O2]tri-1-pyrrolidinylphosphonium hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€120.00
25 G
€303.00

€120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€120.00
25 G
€303.00

About This Item

Empirical Formula (Hill Notation):
C17H29F6N5O3P2
CAS Number:
Molecular Weight:
527.38
MDL number:
UNSPSC Code:
12352126
PubChem Substance ID:
NACRES:
NA.22

€120.00


Check Cart for Availability

Request a Bulk Order

Product Name

PyOxim, 98%

Quality Level

Assay

≥97.5% (HPLC)
98%

form

powder

reaction suitability

reaction type: Coupling Reactions

impurities

≤0.5% water

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CCOC(=O)C(=N\O[P+](N1CCCC1)(N2CCCC2)N3CCCC3)\C#N

InChI

1S/C17H29N5O3P.F6P/c1-2-24-17(23)16(15-18)19-25-26(20-9-3-4-10-20,21-11-5-6-12-21)22-13-7-8-14-22;1-7(2,3,4,5)6/h2-14H2,1H3;/q+1;-1/b19-16+;

InChI key

RDWDVLFMPFUBDV-PXMDEAMVSA-N

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service