Skip to Content
Merck
All Photos(1)

Key Documents

726966

Sigma-Aldrich

(R)-3-Chloro-α-methylbenzylamine

ChiPros®, produced by BASF, ≥99.0%

Synonym(s):

(R)-1-(3-Chlorophenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10ClN
CAS Number:
Molecular Weight:
155.62
Beilstein:
4230572
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

produced by BASF

Assay

≥99.0% (GC)
≥99.0%

form

liquid

optical purity

enantiomeric excess: ≥98.5% (GC)

SMILES string

C[C@@H](N)c1cccc(Cl)c1

InChI

1S/C8H10ClN/c1-6(10)7-3-2-4-8(9)5-7/h2-6H,10H2,1H3/t6-/m1/s1

InChI key

DQEYVZASLGNODG-ZCFIWIBFSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service