Skip to Content
Merck
All Photos(1)

Key Documents

725978

Sigma-Aldrich

1,3-(S)-O-Benzylidene-D-threitol

≥98% (HPLC)

Synonym(s):

(2S,4R,5R)-5-Hydroxy-2-phenyl-1,3-dioxane-4-methanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€192.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€192.00

About This Item

Empirical Formula (Hill Notation):
C11H14O4
CAS Number:
Molecular Weight:
210.23
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€192.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -4.0±0.5°, c = 1 in methanol

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](OC[C@H]1O)c2ccccc2

InChI

1S/C11H14O4/c12-6-10-9(13)7-14-11(15-10)8-4-2-1-3-5-8/h1-5,9-13H,6-7H2/t9-,10-,11+/m1/s1

InChI key

GUOMTMWCUMBRFX-MXWKQRLJSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service