725307
(S)-2-(1-Chloro-2-methyl-2-propanyl)-4-isopropyl-4,5-dihydrooxazole
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H18ClNO
CAS Number:
Molecular Weight:
203.71
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Assay
97%
form
liquid
optical activity
[α]22/D −69.0°, c = 1 in chloroform
refractive index
n20/D 1.491
density
1.009 g/mL at 25 °C
SMILES string
CC(C)[C@H]1COC(=N1)C(C)(C)CCl
InChI
1S/C10H18ClNO/c1-7(2)8-5-13-9(12-8)10(3,4)6-11/h7-8H,5-6H2,1-4H3/t8-/m1/s1
InChI key
KKZCAJJMWXCQPL-MRVPVSSYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service