Skip to Content
Merck
All Photos(1)

Key Documents

725188

Sigma-Aldrich

2-Bromo-3-chloropyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€103.00

About This Item

Empirical Formula (Hill Notation):
C5H3BrClN
CAS Number:
Molecular Weight:
192.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€103.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

58-63 °C

functional group

bromo
chloro

SMILES string

Clc1cccnc1Br

InChI

1S/C5H3BrClN/c6-5-4(7)2-1-3-8-5/h1-3H

InChI key

GOHBBINNYAWQGO-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service