Skip to Content
Merck
All Photos(3)

Key Documents

724637

Sigma-Aldrich

4-Bromoisoxazole

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€90.30

€90.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€90.30

About This Item

Empirical Formula (Hill Notation):
C3H2BrNO
CAS Number:
Molecular Weight:
147.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€90.30


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

mp

38-43 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

Brc1cnoc1

InChI

1S/C3H2BrNO/c4-3-1-5-6-2-3/h1-2H

InChI key

IRDVIGFQMLUJAM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service