724408
1-Butyl-3-methylimidazolium thiocyanate
for energy applications, ≥95%
Synonym(s):
1-Butyl-3-methyl-1H-imidazolium thiocyanate
About This Item
Recommended Products
grade
for energy applications
Assay
≥95%
form
liquid
impurities
0.7% water
refractive index
n20/D 1.538
mp
-28 °C (lit.)
density
1.07 g/mL at 25 °C
anion traces
chloride (Cl-): 0.24%
application(s)
battery manufacturing
SMILES string
[S-]C#N.CCCCn1cc[n+](C)c1
InChI
1S/C8H15N2.CHNS/c1-3-4-5-10-7-6-9(2)8-10;2-1-3/h6-8H,3-5H2,1-2H3;3H/q+1;/p-1
InChI key
SIXHYMZEOJSYQH-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3
Supplementary Hazards
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
403.7 °F - closed cup
Flash Point(C)
206.5 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Ionic liquids, also called room temperature ionic liquids, are organic salts that are liquid at, or close to, room temperature.
Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.
Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.
The critical technical challenges associated with the commercialization of electric vehicle batteries include cost, performance, abuse tolerance, and lifespan.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service