Skip to Content
Merck
All Photos(1)

Key Documents

723401

Sigma-Aldrich

S-Methyl-13C methanethiosulfonate

97 atom % 13C, 97% (CP)

Synonym(s):

S-Methyl-13C thiomethanesulfonate, MMTS-13C

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€322.00

€322.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
10 MG
€322.00

About This Item

Linear Formula:
CH3SO2S13CH3
CAS Number:
Molecular Weight:
127.19
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

€322.00


Estimated to ship on29 May 2025


Request more information

isotopic purity

97 atom % 13C

Quality Level

Assay

97% (CP)

form

liquid

technique(s)

bio NMR: suitable

bp

69-71 °C/0.4 mmHg (lit.)

density

1.347 g/mL at 25 °C

mass shift

M+1

SMILES string

CS(=O)(=O)S[13CH3]

InChI

1S/C2H6O2S2/c1-5-6(2,3)4/h1-2H3/i1+1

InChI key

XYONNSVDNIRXKZ-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service