Skip to Content
Merck
All Photos(1)

Key Documents

722820

Sigma-Aldrich

Ferulic acid-1,2,3-13C3

99 atom % 13C, 98% (CP)

Synonym(s):

trans-4-Hydroxy-3-methoxycinnamic acid-13C3

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€658.00
5 MG
€3,300.00

€658.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
€658.00
5 MG
€3,300.00

About This Item

Linear Formula:
HOC6H3(OCH3)13CH=13CH13CO2H
CAS Number:
Molecular Weight:
197.16
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

€658.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

168-172 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

COc1cc(ccc1O)\[13CH]=[13CH]\[13C](O)=O

InChI

1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+/i3+1,5+1,10+1

InChI key

KSEBMYQBYZTDHS-SQDGEFSZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Bullón-Vela et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-26)
Metabolic syndrome (MetS) components are strongly associated with increased risk of non-alcoholic fatty liver disease (NAFLD) development. Several studies have supported that resveratrol is associated with anti-inflammatory and antioxidant effects on health status. The main objective of this study was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service