Skip to Content
Merck
All Photos(1)

Key Documents

722413

Sigma-Aldrich

2-Bromo-4-chloropyridine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€148.00

€148.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€148.00

About This Item

Empirical Formula (Hill Notation):
C5H3BrClN
CAS Number:
Molecular Weight:
192.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€148.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

mp

12-17 °C

functional group

bromo
chloro

storage temp.

−20°C

SMILES string

Clc1ccnc(Br)c1

InChI

1S/C5H3BrClN/c6-5-3-4(7)1-2-8-5/h1-3H

InChI key

SURKZMFXICWLHU-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service