Skip to Content
Merck
All Photos(1)

Key Documents

722162

Sigma-Aldrich

Cbz-3-Bromopyrrolidine

Synonym(s):

Benzyl 3-bromopyrrolidine-1-carboxylate, Z-3-Bromopyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€84.30

€84.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€84.30

About This Item

Empirical Formula (Hill Notation):
C12H14BrNO2
CAS Number:
Molecular Weight:
284.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€84.30


Check Cart for Availability

Request a Bulk Order

form

liquid

refractive index

n20/D 1.557

density

1.396 g/mL at 25 °C

functional group

bromo
phenyl

storage temp.

2-8°C

SMILES string

BrC1CCN(C1)C(=O)OCc2ccccc2

InChI

1S/C12H14BrNO2/c13-11-6-7-14(8-11)12(15)16-9-10-4-2-1-3-5-10/h1-5,11H,6-9H2

InChI key

LWEKWWHCQBWMQC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service