Skip to Content
Merck
All Photos(1)

Key Documents

722022

Sigma-Aldrich

1-Boc-piperidine-4-carboxaldehyde

95%

Synonym(s):

4-Formylcyclohexanecarboxylic acid tert-butyl ester, tert-Butyl 4-formylpiperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€84.10

€84.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€84.10

About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€84.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

34-40 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCC(CC1)C=O

InChI

1S/C11H19NO3/c1-11(2,3)15-10(14)12-6-4-9(8-13)5-7-12/h8-9H,4-7H2,1-3H3

InChI key

JYUQEWCJWDGCRX-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Pim-1 inhibitors
  • Selective GPR119 agonists for type II diabetes
  • M-tropic (R5) HIV-1 replication inhibitors
  • HDAC inhibitors
  • Selective 5-HT6 antagonists

Reactant for three-component vinylogous Mannich reactions

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service