Skip to Content
Merck
All Photos(1)

Key Documents

720550

Sigma-Aldrich

(3aR,8aR)-4,4,8,8-tetrakis(3,5-di-tert-butylphenyl)-2,2-diethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine

≥94%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€394.00

€394.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€394.00

About This Item

Empirical Formula (Hill Notation):
C71H101O4P
CAS Number:
Molecular Weight:
1049.53
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€394.00


Check Cart for Availability

Request a Bulk Order

Assay

≥94%

form

solid

mp

187-193 °C

functional group

ether
ketal

storage temp.

−20°C

SMILES string

CCC1(CC)O[C@@H]2[C@@H](O1)C(OP(OC2(c3cc(cc(c3)C(C)(C)C)C(C)(C)C)c4cc(cc(c4)C(C)(C)C)C(C)(C)C)c5ccccc5)(c6cc(cc(c6)C(C)(C)C)C(C)(C)C)c7cc(cc(c7)C(C)(C)C)C(C)(C)C

InChI

1S/C71H101O4P/c1-27-69(28-2)72-59-60(73-69)71(56-42-50(65(15,16)17)36-51(43-56)66(18,19)20,57-44-52(67(21,22)23)37-53(45-57)68(24,25)26)75-76(58-32-30-29-31-33-58)74-70(59,54-38-46(61(3,4)5)34-47(39-54)62(6,7)8)55-40-48(63(9,10)11)35-49(41-55)64(12,13)14/h29-45,59-60H,27-28H2,1-26H3/t59-,60-/m1/s1

InChI key

JKEBUWSLWMBNHR-FNSAWIKJSA-N

Application

Reactant for:
  • Preparation of potent and selective small molecule inhibitors of specific isoforms of Cdc2-like kinases (Clk) and dual specificity tyrosine-phosphorylation-regulated kinases (Dyrk)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service