Skip to Content
Merck
All Photos(1)

Key Documents

719706

Sigma-Aldrich

N-Boc-ε-caprolactam

97%

Synonym(s):

1-(tert-Butoxycarbonyl)-1-azacycloheptan-2-one, 1-Boc-2-oxohexamethyleneimine, 1-Boc-aza-2-cycloheptanone, N-tert-Butoxycarbonylhexahydro-2H-azepin-2-one, tert-Butyl-2-oxoazepane-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€104.00

€104.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€104.00

About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€104.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.468

density

1.038 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)N1CCCCCC1=O

InChI

1S/C11H19NO3/c1-11(2,3)15-10(14)12-8-6-4-5-7-9(12)13/h4-8H2,1-3H3

InChI key

GJZYNYJIBDCRFC-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service