Skip to Content
Merck
All Photos(2)

Key Documents

718831

Sigma-Aldrich

4-(Carboxymethyl)phenylboronic acid pinacol ester

95%

Synonym(s):

2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenylacetic acid, Phenylacetic acid-4-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€107.00

€107.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€107.00

About This Item

Empirical Formula (Hill Notation):
C14H19BO4
CAS Number:
Molecular Weight:
262.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€107.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

165-170 °C

functional group

carboxylic acid

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(CC(O)=O)cc2

InChI

1S/C14H19BO4/c1-13(2)14(3,4)19-15(18-13)11-7-5-10(6-8-11)9-12(16)17/h5-8H,9H2,1-4H3,(H,16,17)

InChI key

FNLWHBHWDXCWHV-UHFFFAOYSA-N

Application

4-(Carboxymethyl)phenylboronic acid pinacol ester can be used:
  • As an intermediate to prepare hydantoin-derived autotaxin inhibitors.[1]
  • As a substrate in the synthesis of reactive oxygen species (ROS)-sensitive and H2O2-eliminating materials by interlinking phenylboronic acid pinacol esters onto β-cyclodextrin.[2]
  • To prepare 5-aryl-2-aminopyridine derived FLT3 kinase inhibitors.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a novel class of autotaxin inhibitors through cross-screening
Castagna D, et al.
MedChemComm, 6(6), 1149-1155 (2015)
Structure-property correlations of reactive oxygen species-responsive and hydrogen peroxide-eliminating materials with anti-oxidant and anti-inflammatory activities
Zhang Q, et al.
Chemistry of Materials, 29(19), 8221-8238 (2017)
Discovery and optimization of a highly efficacious class of 5-aryl-2-aminopyridines as FMS-like tyrosine kinase 3 (FLT3) inhibitors
Liu G, et al.
Bioorganic & Medicinal Chemistry Letters, 25(17), 3436-3441 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service