Skip to Content
Merck
All Photos(2)

Key Documents

718386

Sigma-Aldrich

Lithium diisobutyl-tert-butoxyaluminum hydride solution

0.25 M in THF/hexanes

Synonym(s):

LDBBA

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€145.00
100 ML
€398.00

€145.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€145.00
100 ML
€398.00

About This Item

Empirical Formula (Hill Notation):
C12H28AlLiO
CAS Number:
Molecular Weight:
222.27
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€145.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

0.25 M in THF/hexanes

density

0.795 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].CC(C)C[AlH-](CC(C)C)OC(C)(C)C

InChI

1S/C4H9O.2C4H9.Al.Li.H/c1-4(2,3)5;2*1-4(2)3;;;/h1-3H3;2*4H,1H2,2-3H3;;;/q-1;;;;+1;

InChI key

HNIYOHKCLUHKGY-UHFFFAOYSA-N

Application

Lithiumdiisobutyl-tert-butoxyaluminum hydride solution can be used as a reducing agent for the reduction of:
  • Aromatic and aliphatic nitriles to aldehydes.
  • Aromatic and aliphatic esters to aldehydes.
  • Tertiary amides to aldehydes.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service