Skip to Content
Merck
All Photos(2)

Key Documents

717991

Sigma-Aldrich

Lithium thioethoxide

80%

Synonym(s):

Lithium ethanethiolate, Lithium ethyl sulfide, Lithium ethyl thiolate, Lithium ethylmercaptide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€291.00

€291.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€291.00

About This Item

Empirical Formula (Hill Notation):
C2H5LiS
CAS Number:
Molecular Weight:
68.07
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

€291.00


Check Cart for Availability

Request a Bulk Order

Assay

80%

form

solid

mp

316-333 °C

SMILES string

[Li]SCC

InChI

1S/C2H6S.Li/c1-2-3;/h3H,2H2,1H3;/q;+1/p-1

InChI key

JPSNJFMTQIIVFG-UHFFFAOYSA-M

Application

Reactant for preparation of:
  • Microwave-assisted cleavage of aryl methyl ethers
  • Triply thiolate-bridged diiron complexes
  • (+)-batzelladine A and (-)-batzelladine D

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service