Skip to Content
Merck
All Photos(1)

Key Documents

717010

Sigma-Aldrich

N-Boc-cis-4-Fluoro-L-proline methyl ester

97%

Synonym(s):

(2S,4S)-1-tert-Butyl-2-methyl-4-fluoropyrrolidine-1,2-dicarboxylate, Methyl-(2S,4S)-N-Boc-4-fluoropyrrolidine-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€137.00

€137.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€137.00

About This Item

Empirical Formula (Hill Notation):
C11H18FNO4
CAS Number:
Molecular Weight:
247.26
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€137.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

optical activity

[α]22/D -67.0°, c = 0.5 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

refractive index

n20/D 1.448

density

1.153 g/mL at 25 °C

application(s)

peptide synthesis

SMILES string

COC(=O)[C@@H]1C[C@H](F)CN1C(=O)OC(C)(C)C

InChI

1S/C11H18FNO4/c1-11(2,3)17-10(15)13-6-7(12)5-8(13)9(14)16-4/h7-8H,5-6H2,1-4H3/t7-,8-/m0/s1

InChI key

METPQQHVRNLTRX-YUMQZZPRSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service