Skip to Content
Merck
All Photos(1)

Key Documents

716790

Sigma-Aldrich

Dichlorobis(2-(diisopropylphosphino)-ethylamine)ruthenium(II)

97%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€74.60

€74.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€74.60

About This Item

Empirical Formula (Hill Notation):
C16H40Cl2N2P2Ru
CAS Number:
Molecular Weight:
494.43
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€74.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

154-157 °C

SMILES string

Cl[Ru]Cl.CC(C)P(CCN)C(C)C.CC(C)P(CCN)C(C)C

InChI

1S/2C8H20NP.2ClH.Ru/c2*1-7(2)10(6-5-9)8(3)4;;;/h2*7-8H,5-6,9H2,1-4H3;2*1H;/q;;;;+2/p-2

InChI key

HINVYXXIKGEKJI-UHFFFAOYSA-L

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia, W.; Chen , X.; Guo, R.; Sui-Seng, C.; Amoroso, A.; Lough, A. J.; Abdur-Rashid, K.
J. Chem. Soc., Dalton Trans., 8301-8301 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service