Skip to Content
Merck
All Photos(1)

Key Documents

716634

Sigma-Aldrich

(1R,2R)-N,N′-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€101.00

€101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€101.00

About This Item

Empirical Formula (Hill Notation):
C44H44N2P2
CAS Number:
Molecular Weight:
662.78
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€101.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

optical activity

[α]22/D −17.5°, c = 0.5 in chloroform

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

>300 °C

functional group

phosphine

SMILES string

C1CC[C@@H](NCc2ccccc2P(c3ccccc3)c4ccccc4)[C@@H](C1)NCc5ccccc5P(c6ccccc6)c7ccccc7

InChI

1S/C44H44N2P2/c1-5-21-37(22-6-1)47(38-23-7-2-8-24-38)43-31-17-13-19-35(43)33-45-41-29-15-16-30-42(41)46-34-36-20-14-18-32-44(36)48(39-25-9-3-10-26-39)40-27-11-4-12-28-40/h1-14,17-28,31-32,41-42,45-46H,15-16,29-30,33-34H2/t41-,42-/m1/s1

InChI key

OBHPYVNBXWUKNY-NCRNUEESSA-N

Related Categories

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service