Skip to Content
Merck
All Photos(1)

Key Documents

71613

Sigma-Aldrich

O-(2-Azidoethyl)-O-[2-(diglycolyl-amino)ethyl]heptaethylene glycol

≥90% (oligomer purity)

Synonym(s):

Azido-PEG-acid (n=8)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€444.00

€444.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€444.00

About This Item

Empirical Formula (Hill Notation):
C22H42N4O12
CAS Number:
Molecular Weight:
554.59
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€444.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90% (oligomer purity)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

azide
carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(COCC(O)=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C22H42N4O12/c23-26-25-2-4-31-6-8-33-10-12-35-14-16-37-18-17-36-15-13-34-11-9-32-7-5-30-3-1-24-21(27)19-38-20-22(28)29/h1-20H2,(H,24,27)(H,28,29)

InChI key

WWDNBBVPYDZICO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service