Skip to Content
Merck
All Photos(1)

Key Documents

715522

Sigma-Aldrich

(1R,2R)-N,N′-Dimethyl-1,2-diphenylethane-1,2-diamine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€108.00

€108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€108.00

About This Item

Empirical Formula (Hill Notation):
C16H20N2
CAS Number:
Molecular Weight:
240.34
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€108.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

optical activity

[α]22/D +17.0°, c = 0.5 in chloroform

mp

48-53 °C

functional group

amine
phenyl

SMILES string

CN[C@@H]([C@H](NC)c1ccccc1)c2ccccc2

InChI

1S/C16H20N2/c1-17-15(13-9-5-3-6-10-13)16(18-2)14-11-7-4-8-12-14/h3-12,15-18H,1-2H3/t15-,16-/m1/s1

InChI key

BTHYVQUNQHWNLS-HZPDHXFCSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service