Skip to Content
Merck
All Photos(1)

Key Documents

713759

Sigma-Aldrich

1-Bromo-2-ethoxy-benzene

≥97.0% (GC)

Synonym(s):

2-Bromoethoxybenzene, 2-Bromophenetole, o-Bromophenetole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrO
CAS Number:
Molecular Weight:
201.06
Beilstein:
2043678
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

SMILES string

CCOc1ccccc1Br

InChI

1S/C8H9BrO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2H2,1H3

InChI key

JVEQWIQHHWNMQX-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service