Skip to Content
Merck
All Photos(2)

Key Documents

713724

Sigma-Aldrich

Ethyl N-Boc-piperidine-3-carboxylate

≥97.0%

Synonym(s):

1-(tert-Butoxycarbonyl)-piperidine-3-carboxylic acid ethyl ester, 1-(tert-Butyl) 3-ethyl 1,3-piperidinedicarboxylate, N-Boc-DL-nipecotic acid ethyl ester, Ethyl N-Boc-DL-nipecotate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H23NO4
CAS Number:
Molecular Weight:
257.33
Beilstein:
3614917
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)
≥97.0%

form

powder

SMILES string

CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C

InChI

1S/C13H23NO4/c1-5-17-11(15)10-7-6-8-14(9-10)12(16)18-13(2,3)4/h10H,5-9H2,1-4H3

InChI key

YCXCRFGBFZTUSU-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • GABAA receptor agonists
  • Piperidine derivatives
  • Selective TACE inhibitors
  • HDL-elevating agents
  • α-Sulfonyl hydroxamic acid derivatives
  • chemicals in the Iboga-alkaloid family

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a Novel Heterospirocyclic 3-(N-Methyl-N-phenylamino)-2H-azirine and its use as an Amino Acid Equivalent in the Preparation of a Model Tripeptide
Villalgordo, J.M. and Heimgartner, M.
Tetrahedron Asymmetry, 49 (33), 7215-7222 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service