Skip to Content
Merck
All Photos(1)

Key Documents

713201

Sigma-Aldrich

(8α,9S)-6′-Methoxycinchonan-9-amine trihydrochloride

90%

Synonym(s):

(8S,9S)-9-Amino-9-deoxyepiquinine trihydrochloride, (S)-(6-Methoxyquinolin-4-yl)((2S,4S,5R)-5-vinylquinuclidin-2-yl)methanamine trihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€141.00

€141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€141.00

About This Item

Empirical Formula (Hill Notation):
C20H25N3O · 3HCl
CAS Number:
Molecular Weight:
432.81
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€141.00


Check Cart for Availability

Request a Bulk Order

Assay

89.5-110.5% (AT)
90% (HPLC)
90%

form

powder

optical activity

[α]/D +65.0±2.0°, c = 1 in 0.1 M HCl

functional group

amine

SMILES string

Cl.Cl.Cl.COc1ccc2nccc([C@H](N)C3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H25N3O.3ClH/c1-3-13-12-23-9-7-14(13)10-19(23)20(21)16-6-8-22-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20H,1,7,9-10,12,21H2,2H3;3*1H/t13-,14-,19?,20-;;;/m0.../s1

InChI key

GZXDEKGNTLNSGM-YPIZWHDUSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service