Skip to Content
Merck
All Photos(2)

Key Documents

711438

Sigma-Aldrich

4,6,6-Trimethyl-1,3,2-dioxaborinane solution

1.0 M in dioxane

Synonym(s):

MPBH, Methylpentanediolborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13BO2
Molecular Weight:
127.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

concentration

1.0 M in dioxane

density

1.008 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC1CC(C)(C)OBO1

InChI

1S/C6H13BO2/c1-5-4-6(2,3)9-7-8-5/h5,7H,4H2,1-3H3

InChI key

ARSUQHZDPBOKDP-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Fan et al.
Materials science & engineering. C, Materials for biological applications, 56, 311-317 (2015-08-08)
A robust synthetic strategy of biopolymer-based hydrogels has been developed where hyaluronic acid derivatives reacted through aqueous Diels-Alder chemistry without the involvement of chemical catalysts, allowing for control and sustain release of dexamethasone. To conjugate the hydrogel, furan and maleimide
Wendy Wj Unger et al.
Oncoimmunology, 4(8), e970462-e970462 (2015-09-26)
Therapeutic vaccinations against cancer are still largely ineffective. Major caveats are inefficient delivery of tumor antigens to dendritic cells (DCs) and excessive immune suppression by
Toshiki Matsui et al.
Acta biomaterialia, 13, 32-41 (2014-12-03)
Polylactic acid (PLA) is a candidate material to prepare scaffolds for 3-D tissue regeneration. However, cells do not adhere or proliferate well on the surface of PLA because it is hydrophobic. We report a simple and rapid method for inducing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service