Skip to Content
Merck
All Photos(1)

Key Documents

710881

Sigma-Aldrich

Potassium phenoxymethyltrifluoroborate

contains 20% KBr, 97%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€90.00

€90.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€90.00

About This Item

Empirical Formula (Hill Notation):
C7H7BF3KO
CAS Number:
Molecular Weight:
214.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€90.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

contains

20% KBr

mp

>300 °C

functional group

phenoxy

SMILES string

[K+].F[B-](F)(F)COc1ccccc1

InChI

1S/C7H7BF3O.K/c9-8(10,11)6-12-7-4-2-1-3-5-7;/h1-5H,6H2;/q-1;+1

InChI key

OVYSNXCRBZPJIG-UHFFFAOYSA-N

General description

Potassium phenoxymethyltrifluoroborate is a stable boronic acid surrogate useful for Suzuki-Miyaura cross-coupling reactions and other C-C bond forming reactions.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aryloxymethyltrifluoroborates for rhodium-catalyzed asymmetric conjugate Arylation. o-methoxyarylation through 1, 4-rhodium shift
Ming J and Hayashi T
Organic Letters, 18(24), 6452-6455 (2016)
Preparation of potassium alkoxymethyltrifluoroborates and their cross-coupling with aryl chlorides
Molander GA and Canturk B
Organic Letters, 10(11), 2135-2138 (2008)

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service