All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
liquid
refractive index
n20/D 1.499
density
0.995 g/mL at 25 °C
SMILES string
COC(Cc1ccccc1C)OC
InChI
1S/C11H16O2/c1-9-6-4-5-7-10(9)8-11(12-2)13-3/h4-7,11H,8H2,1-3H3
InChI key
LQXUNLBAEJOILA-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service