Skip to Content
Merck
All Photos(1)

Key Documents

709182

Sigma-Aldrich

Molybdenum boride

powder, −325 mesh, 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€114.00

€114.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€114.00

About This Item

Linear Formula:
MoB
CAS Number:
Molecular Weight:
106.75
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

€114.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

99.5% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: boron

reagent type: catalyst
core: molybdenum

particle size

−325 mesh

mp

2180 °C

density

8.65 g/cm3 at 25 °C

SMILES string

B#[Mo]

InChI

1S/B.Mo

InChI key

LGLOITKZTDVGOE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service