Skip to Content
Merck
All Photos(2)

Documents

707473

Sigma-Aldrich

Small gap fullerene-ethyl nipecotate

greener alternative

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C60(C8H15NO2)x
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥95%

form

solid

mol wt

average Mv ~2100

composition

fullerenes, 50%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

~5% o-Dichlorobenzene

mp

>300 °C

solubility

organic solvents: soluble
toluene: soluble

greener alternative category

General description

Insoluble small gap fullerenes can be extracted into organic solvents by simple chemical redox process.
Small gap fullerene-ethyl nipecotate are higher order fullerenes with low HOMO-LUMO gaps. These allotropes of carbon are also known as metallofullerenes. They are kinetically unstable and are considered as insoluble small band gap structures. Small gap fullerene-ethyl nipecotate can be extracted into organic solvents by a chemical redox processes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Legal Information

Product of TDA Research, Inc. and US Patents 6,517,799 and 6,303,016.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical redox recovery of giant, small-gap and other fullerenes
Raebiger, J.W.; Alford, J.M.; Bolskar, R.D.; Diener, M.D.
Carbon, 49, 37-46 (2011)
Isolation and properties of small-bandgap fullerenes.
Diener MD and Alford JM
Nature, 393(6686), 668-668 (1998)
Small-bandgap endohedral metallofullerenes in high yield and purity.
Stevenson S, et al.
Nature, 401(6748), 55-55 (1999)

Articles

Find various photovoltaic and bioscience-based applications of fullerenes.

SWCNTs show promise in FETs, solar cells, and photodetectors due to their ultrafast charge transport mobility.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service