706892
Methyl 2-bromopyridine-3-carboxylate
97%
Synonym(s):
2-Bromonicotinic acid methyl ester, Methyl 2-bromonicotinate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6BrNO2
CAS Number:
Molecular Weight:
216.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
solid
mp
32-37 °C
SMILES string
COC(=O)c1cccnc1Br
InChI
1S/C7H6BrNO2/c1-11-7(10)5-3-2-4-9-6(5)8/h2-4H,1H3
InChI key
RAFFKXWNTXTTFO-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service