Skip to Content
Merck
All Photos(1)

Key Documents

706531

Sigma-Aldrich

N-Boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

95%

Synonym(s):

(1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridin-4-yl)boronic acid pinacol ester, (N-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridin-4-yl)boronic acid pinacol ester, (N-tert-Butoxycarbonyl-1,2,5,6-tetrahydropyridin-4-yl)boronic acid pinacol ester, (N-tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€130.00

€130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€130.00

About This Item

Empirical Formula (Hill Notation):
C16H28BNO4
CAS Number:
Molecular Weight:
309.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€130.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

mp

100-114 °C

SMILES string

CC(C)(C)OC(=O)N1CCC(=CC1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C16H28BNO4/c1-14(2,3)20-13(19)18-10-8-12(9-11-18)17-21-15(4,5)16(6,7)22-17/h8H,9-11H2,1-7H3

InChI key

VVDCRJGWILREQH-UHFFFAOYSA-N

General description

Boc-THP-Bpin is frequently used in metal catalyzed cross-coupling reactions, such as Suzuki-Miyaura and Negishi couplings.[1][2]

Application

Reagent used for
  • Suzuki-Miyaura cross-coupling using palladium phosphine catalyst[1]
  • Palladium-catalyzed ligand-controlled regioselective Suzuki coupling[2]
  • Palladium-catalyzed Suzuki-Miyaura coupling[3]
  • Suzuki coupling followed by iodolactonization reaction[4]
  • Wrenchnolol derivative optimized for gene activation in cells[5]

Reagent used in Preparation of several enzymatic inhibitors and receptor ligands
  • Orally active anaplastic lymphoma kinase inhibitors[6]
  • Oxazolecarboxamides as diacylglycerol acyltransferase-1 inhibitors for treatment of obesity and diabetes[7]
  • 4-arylpiperidinyl amides and N-arylpiperidin-3-yl-cyclopropanecarboxamides as novel melatonin receptor ligands[8]
  • Quinazoline analogs as glucocerebrosidase inhibitors with chaperone activity for treatment of Gaucher disease, a lysosomal storage disorder[9]
  • Arylpiperazine and piperidine ethers as dual acting norepinephrine reuptake inhibitors and 5-HT1A partial agonists[10]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of 3,4-fused-spiro[furan-5(5H),4'-piperidin]-2-one
Liu, J.; et al.
Tetrahedron Letters, 50, 5228-5230 (2009)
Kazutomo Kinoshita et al.
Bioorganic & medicinal chemistry, 20(3), 1271-1280 (2012-01-10)
Anaplastic lymphoma kinase (ALK) receptor tyrosine kinase is considered an attractive therapeutic target for human cancers, especially non-small cell lung cancer (NSCLC). Our previous study revealed that 8,9-side-chains of 6,6-dimethyl-11-oxo-6,11-dihydro-5H-benzo[b]carbazole scaffold crucially affected kinase selectivity, cellular activity, and metabolic stability.
Yimin Qian et al.
Journal of medicinal chemistry, 54(7), 2433-2446 (2011-03-19)
Diacylglycerol acyltransferase-1 (DGAT-1) is the enzyme that catalyzes the final and committed step of triglyceride formation, namely, the acylation of diacylglycerol with acyl coenzyme A. DGAT-1 deficient mice demonstrate resistance to weight gain on high fat diet, improved insulin sensitivity
Discovery and pharmacological characterization of aryl piperazine and piperidine ethers as dual acting norepinephrine reuptake inhibitors and 5-HT1A partial agonists
Gray, D. L.; et al.
Bioorganic & Medicinal Chemistry, 19, 6604-6607 (2009)
Juan J Marugan et al.
Journal of medicinal chemistry, 54(4), 1033-1058 (2011-01-22)
Gaucher disease is a lysosomal storage disorder (LSD) caused by deficiency in the enzyme glucocerebrosidase (GC). Small molecule chaperones of protein folding and translocation have been proposed as a promising therapeutic approach to this LSD. Most small molecule chaperones described

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service